Learn More
3-Benzoylbenzoic acid, 98%, Thermo Scientific Chemicals
$98.58 - $380.04
Chemical Identifiers
CAS | 579-18-0 |
---|---|
Molecular Formula | C14H10O3 |
Molecular Weight (g/mol) | 226.23 |
MDL Number | MFCD00002518 |
InChI Key | AXJXRLHTQQONQR-UHFFFAOYSA-N |
PubChem CID | 101386 |
IUPAC Name | 3-benzoylbenzoic acid |
SMILES | OC(=O)C1=CC=CC(=C1)C(=O)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2551906
|
Thermo Scientific Chemicals
B2551906 |
5 g |
Each for $98.58
|
|
|||||
AAB2551914
|
Thermo Scientific Chemicals
B2551914 |
25 g |
Each for $380.04
|
|
|||||
Description
3-Benzoylbenzoic acid has been used in light-induced reactions on benzophenone-terminated boron-doped diamond (BDD) surfaces and preparation of benzophenone flavonol derivative.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Benzoylbenzoic acid has been used in light-induced reactions on benzophenone-terminated boron-doped diamond (BDD) surfaces and preparation of benzophenone flavonol derivative.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
579-18-0 | |
226.23 | |
AXJXRLHTQQONQR-UHFFFAOYSA-N | |
3-benzoylbenzoic acid |
C14H10O3 | |
MFCD00002518 | |
101386 | |
OC(=O)C1=CC=CC(=C1)C(=O)C1=CC=CC=C1 |
Specifications
579-18-0 | |
Odorless | |
MFCD00002518 | |
2212366 | |
Insoluble in water. | |
OC(=O)C1=CC=CC(=C1)C(=O)C1=CC=CC=C1 | |
226.23 | |
226.23 | |
3-Benzoylbenzoic acid |
162°C to 166°C | |
C14H10O3 | |
5 g | |
benzoic acid, 3-benzoyl, m-benzoylbenzoic acid, 3-benzoyl benzoic acid, 3-benzoyl-benzoic acid, 3-phenylcarbonyl benzoic acid, 3-benzoylbenzoicacid, 3-benzoyl-benzoesaure, 3-benzoylbenzoic-acid, acmc-20an9k, benzoic acid,3-benzoyl | |
AXJXRLHTQQONQR-UHFFFAOYSA-N | |
3-benzoylbenzoic acid | |
101386 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only