Learn More
3-(2-Furyl)acrolein, 99%, Thermo Scientific Chemicals
$55.05 - $182.42
Chemical Identifiers
CAS | 623-30-3 |
---|---|
Molecular Formula | C7H6O2 |
Molecular Weight (g/mol) | 122.123 |
MDL Number | MFCD00003256 |
InChI Key | VZIRCHXYMBFNFD-HNQUOIGGSA-N |
Synonym | 3-2-furyl acrolein, 2-furanacrolein, e-3-furan-2-yl acrylaldehyde, 3-furan-2-yl acrylaldehyde, 2-propenal, 3-2-furanyl, trans-3-2-furyl acrolein, beta-2-furylacrolein, 3-2-furyl acrylaldehyde, unii-ssq86u3drp, furyl acrolein |
PubChem CID | 1549521 |
IUPAC Name | (E)-3-(furan-2-yl)prop-2-enal |
SMILES | C1=COC(=C1)C=CC=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1304206
|
Thermo Scientific Chemicals
A1304206 |
5 g |
Each for $55.05
|
|
|||||
AAA1304214
|
Thermo Scientific Chemicals
A1304214 |
25 g |
Each for $182.42
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
623-30-3 | |
122.123 | |
VZIRCHXYMBFNFD-HNQUOIGGSA-N | |
1549521 | |
C1=COC(=C1)C=CC=O |
C7H6O2 | |
MFCD00003256 | |
3-2-furyl acrolein, 2-furanacrolein, e-3-furan-2-yl acrylaldehyde, 3-furan-2-yl acrylaldehyde, 2-propenal, 3-2-furanyl, trans-3-2-furyl acrolein, beta-2-furylacrolein, 3-2-furyl acrylaldehyde, unii-ssq86u3drp, furyl acrolein | |
(E)-3-(furan-2-yl)prop-2-enal |
Specifications
623-30-3 | |
95°C (5 mmHg) | |
C7H6O2 | |
5 g | |
107570 | |
3-2-furyl acrolein, 2-furanacrolein, e-3-furan-2-yl acrylaldehyde, 3-furan-2-yl acrylaldehyde, 2-propenal, 3-2-furanyl, trans-3-2-furyl acrolein, beta-2-furylacrolein, 3-2-furyl acrylaldehyde, unii-ssq86u3drp, furyl acrolein | |
C1=COC(=C1)C=CC=O | |
122.123 | |
122.12 | |
3-(2-Furyl)acrolein |
49°C to 52°C | |
99°C (210°F) | |
MFCD00003256 | |
UN1759 | |
Air sensitive | |
VZIRCHXYMBFNFD-HNQUOIGGSA-N | |
(E)-3-(furan-2-yl)prop-2-enal | |
1549521 | |
99% |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber : 210-785-3
RTECSNumber : LT8528500
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only