missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-[2-(4-Morpholinyl)ethylcarbamoyl]benzeneboronic acid hydrochloride, 98%, Thermo Scientific™

$145.32 - $145.32

Chemical Identifiers

CAS 957060-89-8
Molecular Formula C13H20BClN2O4
Molecular Weight (g/mol) 314.573
MDL Number MFCD09027255
InChI Key QBUJRGQOXYFCFK-UHFFFAOYSA-N
Synonym 3-2-morpholinoethyl carbamoyl phenyl boronic acid hydrochloride, 3-2-morpholin-4-ylethyl carbamoyl benzeneboronic acid hydrochloride, 3-2-morpholin-4-yl ethyl carbamoyl phenylboronic acid hydrochloride, 3-2-morpholinoethylcarbamoyl phenylboronic acid hcl, 3-2-morpholinoethylcarbamoyl phenylboronic acid, hcl, 3-2-morpholinoethyl carbamoyl phenylboronic acid hydrochloride, 3-2-morpholinoethyl carbamoyl phenyl boronicacidhydrochloride, 3-2-morpholin-4-yl ethyl carbamoyl phenyl boronic acid-hydrogen chloride 1/1
PubChem CID 44119393
IUPAC Name [3-(2-morpholin-4-ylethylcarbamoyl)phenyl]boronic acid;hydrochloride
SMILES B(C1=CC(=CC=C1)C(=O)NCCN2CCOCC2)(O)O.Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH52689MD
View Documents Promotion Details
Thermo Scientific Chemicals
H52689MD
250 mg
Each for $145.32
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Insoluble in water. Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

957060-89-8
314.573
QBUJRGQOXYFCFK-UHFFFAOYSA-N
44119393
B(C1=CC(=CC=C1)C(=O)NCCN2CCOCC2)(O)O.Cl
C13H20BClN2O4
MFCD09027255
3-2-morpholinoethyl carbamoyl phenyl boronic acid hydrochloride, 3-2-morpholin-4-ylethyl carbamoyl benzeneboronic acid hydrochloride, 3-2-morpholin-4-yl ethyl carbamoyl phenylboronic acid hydrochloride, 3-2-morpholinoethylcarbamoyl phenylboronic acid hcl, 3-2-morpholinoethylcarbamoyl phenylboronic acid, hcl, 3-2-morpholinoethyl carbamoyl phenylboronic acid hydrochloride, 3-2-morpholinoethyl carbamoyl phenyl boronicacidhydrochloride, 3-2-morpholin-4-yl ethyl carbamoyl phenyl boronic acid-hydrogen chloride 1/1
[3-(2-morpholin-4-ylethylcarbamoyl)phenyl]boronic acid;hydrochloride

Specifications

957060-89-8
C13H20BClN2O4
250 mg
Insoluble in water. Soluble in organic solvents.
B(C1=CC(=CC=C1)C(=O)NCCN2CCOCC2)(O)O.Cl
314.573
314.58
3-[2-(4-Morpholinyl)ethylcarbamoyl]benzeneboronic acid hydrochloride
190°C to 195°C
MFCD09027255
3-2-morpholinoethyl carbamoyl phenyl boronic acid hydrochloride, 3-2-morpholin-4-ylethyl carbamoyl benzeneboronic acid hydrochloride, 3-2-morpholin-4-yl ethyl carbamoyl phenylboronic acid hydrochloride, 3-2-morpholinoethylcarbamoyl phenylboronic acid hcl, 3-2-morpholinoethylcarbamoyl phenylboronic acid, hcl, 3-2-morpholinoethyl carbamoyl phenylboronic acid hydrochloride, 3-2-morpholinoethyl carbamoyl phenyl boronicacidhydrochloride, 3-2-morpholin-4-yl ethyl carbamoyl phenyl boronic acid-hydrogen chloride 1/1
QBUJRGQOXYFCFK-UHFFFAOYSA-N
[3-(2-morpholin-4-ylethylcarbamoyl)phenyl]boronic acid;hydrochloride
44119393
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'