missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Pyrrolidinone, 99%, Acros Organics™

$33.02 - $55.26

Chemical Identifiers

CAS 616-45-5
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.106
MDL Number MFCD00005270
InChI Key HNJBEVLQSNELDL-UHFFFAOYSA-N
Synonym 2-pyrrolidinone, pyrrolidone, butyrolactam, 2-pyrrolidone, 2-oxopyrrolidine, pyrrolidon, 2-ketopyrrolidine, 2-pyrol, pyrrolidinone, gamma-butyrolactam
PubChem CID 12025
ChEBI CHEBI:36592
IUPAC Name pyrrolidin-2-one
SMILES C1CC(=O)NC1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC132115000 SDS View Documents Acros Organics
132115000
500mL Glass bottle Each for $55.26
AC132112500 SDS View Documents Acros Organics
132112500
250mL Glass bottle Each for $33.02
AC132110025 SDS View Documents Acros Organics
AC132110025
2.5L Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

616-45-5
85.106
HNJBEVLQSNELDL-UHFFFAOYSA-N
12025
pyrrolidin-2-one
C4H7NO
MFCD00005270
2-pyrrolidinone, pyrrolidone, butyrolactam, 2-pyrrolidone, 2-oxopyrrolidine, pyrrolidon, 2-ketopyrrolidine, 2-pyrol, pyrrolidinone, gamma-butyrolactam
CHEBI:36592
C1CC(=O)NC1

Specifications

1.1030g/mL
Authentic
1.4860 to 1.4880
250.0°C
25.5°C
616-45-5
100.0
C4H7NO
21, 236
2-pyrrolidinone, pyrrolidone, butyrolactam, 2-pyrrolidone, 2-oxopyrrolidine, pyrrolidon, 2-ketopyrrolidine, 2-pyrol, pyrrolidinone, gamma-butyrolactam
HNJBEVLQSNELDL-UHFFFAOYSA-N
pyrrolidin-2-one
12025
85.11
99%
138°C
Glass bottle
1.103
Colorless to Yellow
500mL
98.5
99%
MFCD00005270
15, 8130
Solubility in water: miscible. Other solubilities: miscible with ethanol, ether, chloroform, benzene,, ethyl acetate, carbon disulfide
C1CC(=O)NC1
85.106
CHEBI:36592
Liquid or Low Melting Mass
2-Pyrrolidinone, 98%
Safety and Handling

Safety and Handling

GHS P Statement Causes serious eye irritation.

Warning

EINECSNumber : 210-483-1

RTECSNumber : UY5715000

TSCA : TSCA

Recommended Storage : Product may darken during storage