Learn More
Isopropanol, Optima™LC/MS Grade, Fisher Chemical™
2-Propanol, >-99.90%, C3H8O, CAS Number-67-63-0, petrohol, isopropyl alcohol, isopropanol, sec-propyl alcohol, dimethylcarbinol, 2-hydroxypropane, hartosol, 2-propanol, alkolave, avantine, 4L, Report AU at 254nm, Report AU at 210nm, Report AU at 230nm, Report AU at 220nm
$253.25 - $710.75
Chemical Identifiers
CAS | 67-63-0 |
---|---|
Molecular Formula | C3H8O |
Molecular Weight (g/mol) | 60.096 |
MDL Number | 11674 |
InChI Key | KFZMGEQAYNKOFK-UHFFFAOYSA-N |
Synonym | isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol |
PubChem CID | 3776 |
ChEBI | CHEBI:17824 |
IUPAC Name | propan-2-ol |
SMILES | CC(C)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
A461500
|
Fisher Chemical
A461500 |
500 mL | Glass Bottle |
Each for $253.25
Case of 6 Each for $1,519.50
|
|
||||
A4611
|
Fisher Chemical
A4611 |
1 L | Wide Neck Amber Bottle |
Each for $333.42
Case of 6 Each for $2,000.52
|
|
||||
A461212
|
Fisher Chemical
A461212 |
2.5 L | Glass Bottle |
Each for $680.67
Case of 4 Each for $2,722.68
|
|
||||
A4614
|
Fisher Chemical
A4614 |
4 L | Glass Bottle |
Each for $710.75
Case of 4 Each for $2,843.00
|
|
||||
Description
2-Propanol, >-99.90%, C3H8O, CAS Number-67-63-0, petrohol, isopropyl alcohol, isopropanol, sec-propyl alcohol, dimethylcarbinol, 2-hydroxypropane, hartosol, 2-propanol, alkolave, avantine, 4L, Report AU at 254nm, Report AU at 210nm, Report AU at 230nm, Report AU at 220nm
Chemical Identifiers
67-63-0 | |
60.096 | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
3776 | |
propan-2-ol |
C3H8O | |
11674 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CHEBI:17824 | |
CC(C)O |
Specifications
7 | |
67-63-0 | |
Colorless | |
81°C | |
Glass Bottle | |
1ppm max. | |
C3H8O | |
11674 | |
2.27 mPaS at 20°C | |
15, 5254 | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
propan-2-ol | |
3776 | |
60.1 | |
LC-MS & UHPLC-UV |
-89.5°C | |
Report AU at 210nm, Report AU at 220nm, Report AU at 230nm, Report AU at 254nm | |
0.785g/cm³ | |
500 mL | |
Liquid | |
Assay: ≥99.90% | |
(CH3)2CHOH | |
43mmHg at 20°C | |
0.05% max. | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CC(C)O | |
60.096 | |
CHEBI:17824 | |
≥99.90% | |
2-Propanol |
Safety and Handling
WARNING!
Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.
NFPA
Health:1
Flammability:3
Instability:0