missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isopropanol (IPA), Optima™ for HPLC, Fisher Chemical™

High purity mobile phase for HPLC and extraction solvent

Supplier:  Thermo Fisher Scientific A464RS200

 View more versions of this product

Catalog No. A464RS200

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

High purity mobile phase for HPLC and extraction solvent

  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Also meets ACS specifications
  • Intended for research use only
Specifications

Specifications

7
-89.5°C
0.015 AU max. at 254nm, 0.10 AU max. at 230nm, 0.20 AU max. at 220nm, 1.00 AU max. at 205nm
0.785g/cm³
200 L
FisherPak (Rented)
High purity mobile phase for HPLC and/or extraction solvent
99.9% min by GC
(CH3)2CHOH
43mmHg at 20°C
0.05% max.
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
3776
60.01
OPTIMA™
2-Propano
67-63-0
Colorless
81°C
Pass Test
Liquid
1ppm max.
C3H8O
MFCD00011674
2.27 mPa/s at 20°C
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CC(C)O
60.096
CHEBI:17824
≥99.9%

Chemical Identifiers

67-63-0
60.096
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
propan-2-ol
C3H8O
MFCD00011674
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CHEBI:17824
CC(C)O
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and if s. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:1
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

SDS Documents

Documents

Promotions

Promotions