Learn More
(+/-)-2-Methyl-1-butanol, 98%, Thermo Scientific Chemicals
$43.53 - $218.12
Chemical Identifiers
CAS | 137-32-6 |
---|---|
Molecular Formula | C5H12O |
Molecular Weight (g/mol) | 88.15 |
MDL Number | MFCD00004743 |
InChI Key | QPRQEDXDYOZYLA-UHFFFAOYSA-N |
Synonym | 2-methyl-1-butanol, active amyl alcohol, 1-butanol, 2-methyl, 2-methylbutanol, sec-butylcarbinol, dl-2-methyl-1-butanol, 2-methyl-n-butanol, 2-methylbutyl alcohol, 2-methyl butanol-1, primary active amyl alcohol |
PubChem CID | 8723 |
ChEBI | CHEBI:48945 |
IUPAC Name | 2-methylbutan-1-ol |
SMILES | CCC(C)CO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB21825AE
|
Thermo Scientific Chemicals
B21825AE |
100 mL |
Each for $43.53
|
|
|||||
AAB21825AP
|
Thermo Scientific Chemicals
B21825AP |
500 mL |
Each for $69.33
|
|
|||||
AAB218250F
|
Thermo Scientific Chemicals
B218250F |
2500 mL |
Each for $218.12
|
|
|||||
Description
2-Methyl-1-butanol is a volatile metabolite produced by a number of different plant species. Used as a solvent in organic synthesis (introduction of active amyl group), in lubricants, plasticizers, additives for oils & paints. It is also employed as a perfuming agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Methyl-1-butanol is a volatile metabolite produced by a number of different plant species. Used as a solvent in organic synthesis (introduction of active amyl group), in lubricants, plasticizers, additives for oils & paints. It is also employed as a perfuming agent.
Solubility
Soluble in water. [183 g/L (0 C)] and alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
137-32-6 | |
88.15 | |
QPRQEDXDYOZYLA-UHFFFAOYSA-N | |
8723 | |
2-methylbutan-1-ol |
C5H12O | |
MFCD00004743 | |
2-methyl-1-butanol, active amyl alcohol, 1-butanol, 2-methyl, 2-methylbutanol, sec-butylcarbinol, dl-2-methyl-1-butanol, 2-methyl-n-butanol, 2-methylbutyl alcohol, 2-methyl butanol-1, primary active amyl alcohol | |
CHEBI:48945 | |
CCC(C)CO |
Specifications
137-32-6 | |
0.815 | |
43°C (109°F) | |
1.411 | |
100 mL | |
1718810 | |
2-methyl-1-butanol, active amyl alcohol, 1-butanol, 2-methyl, 2-methylbutanol, sec-butylcarbinol, dl-2-methyl-1-butanol, 2-methyl-n-butanol, 2-methylbutyl alcohol, 2-methyl butanol-1, primary active amyl alcohol | |
QPRQEDXDYOZYLA-UHFFFAOYSA-N | |
2-methylbutan-1-ol | |
8723 | |
88.15 | |
(+/-)-2-Methyl-1-butanol |
<-70°C | |
129°C to 131°C | |
C5H12O | |
MFCD00004743 | |
UN1105 | |
14,6030 | |
Soluble in water. [183g/L (0 C)] and alcohol. | |
CCC(C)CO | |
88.15 | |
CHEBI:48945 | |
98% |
Safety and Handling
GHS H Statement
H226-H332-H335
Flammable liquid and vapour.
Harmful if inhaled.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H332-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PENTANOLS
EINECSNumber : 205-289-9
RTECSNumber : EL5250000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only