Learn More
2-Methoxypyridine-3-boronic acid, 98%, Thermo Scientific Chemicals
$140.51 - $439.70
Chemical Identifiers
CAS | 163105-90-6 |
---|---|
Molecular Formula | C6H8BNO3 |
Molecular Weight (g/mol) | 152.94 |
MDL Number | MFCD03411572 |
InChI Key | NVOLYUXUHWBCRJ-UHFFFAOYSA-N |
Synonym | 2-methoxypyridine-3-boronic acid, 2-methoxy-3-pyridineboronic acid, 2-methoxypyridin-3-yl boronic acid, 2-methoxy-3-pyridylboronic acid, 2-methoxypyridin-3-yl-3-boronic acid, 2-methoxy-3-pyridinyl boronic acid, 2-methoxy-3-pyridyl boronic acid, 2-methoxy-3-pyridinyl-boronic acid, boronic acid, 2-methoxy-3-pyridinyl, boronic acid, b-2-methoxy-3-pyridinyl |
PubChem CID | 2762709 |
IUPAC Name | (2-methoxypyridin-3-yl)boronic acid |
SMILES | COC1=NC=CC=C1B(O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL2009403
|
Thermo Scientific Chemicals
L2009403 |
1 g |
Each for $140.51
|
|
|||||
AAL2009406
|
Thermo Scientific Chemicals
L2009406 |
5 g |
Each for $439.70
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
163105-90-6 | |
152.94 | |
NVOLYUXUHWBCRJ-UHFFFAOYSA-N | |
2762709 | |
COC1=NC=CC=C1B(O)O |
C6H8BNO3 | |
MFCD03411572 | |
2-methoxypyridine-3-boronic acid, 2-methoxy-3-pyridineboronic acid, 2-methoxypyridin-3-yl boronic acid, 2-methoxy-3-pyridylboronic acid, 2-methoxypyridin-3-yl-3-boronic acid, 2-methoxy-3-pyridinyl boronic acid, 2-methoxy-3-pyridyl boronic acid, 2-methoxy-3-pyridinyl-boronic acid, boronic acid, 2-methoxy-3-pyridinyl, boronic acid, b-2-methoxy-3-pyridinyl | |
(2-methoxypyridin-3-yl)boronic acid |
Specifications
163105-90-6 | |
C6H8BNO3 | |
1 g | |
NVOLYUXUHWBCRJ-UHFFFAOYSA-N | |
(2-methoxypyridin-3-yl)boronic acid | |
2762709 | |
98% |
136°C to 138°C | |
MFCD03411572 | |
2-methoxypyridine-3-boronic acid, 2-methoxy-3-pyridineboronic acid, 2-methoxypyridin-3-yl boronic acid, 2-methoxy-3-pyridylboronic acid, 2-methoxypyridin-3-yl-3-boronic acid, 2-methoxy-3-pyridinyl boronic acid, 2-methoxy-3-pyridyl boronic acid, 2-methoxy-3-pyridinyl-boronic acid, boronic acid, 2-methoxy-3-pyridinyl, boronic acid, b-2-methoxy-3-pyridinyl | |
COC1=NC=CC=C1B(O)O | |
152.94 | |
152.95 | |
2-Methoxypyridine-3-boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only