Learn More
2-Iodobutane, 99%, stab. with copper, Thermo Scientific Chemicals
$71.86 - $433.01
Chemical Identifiers
CAS | 513-48-4 |
---|---|
Molecular Formula | C4H9I |
Molecular Weight (g/mol) | 184.02 |
MDL Number | MFCD00001072 |
InChI Key | IQRUSQUYPCHEKN-UHFFFAOYNA-N |
Synonym | sec-butyl iodide, butane, 2-iodo, 2-butyl iodide, sec-iodobutane, 2-jodbutan, 2-jodbutan czech, 2-iodanylbutane, 2-iodobutan, 2-iodo-butane, sec.-butyl iodide |
PubChem CID | 10559 |
IUPAC Name | 2-iodobutane |
SMILES | CCC(C)I |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2061918
|
Thermo Scientific Chemicals
B2061918 |
50 g |
N/A
|
|
|||||
AAB2061930
|
Thermo Scientific Chemicals
B2061930 |
250 g |
N/A
|
|
|||||
AAB2061936
|
Thermo Scientific Chemicals
B2061936 |
500 g |
N/A
|
|
|||||
Description
It is used as a solvent organic synthesis and as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a solvent organic synthesis and as pharmaceutical intermediate.
Solubility
Insoluble in water. Soluble in alcohol and diethyl ether.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Chemical Identifiers
513-48-4 | |
184.02 | |
IQRUSQUYPCHEKN-UHFFFAOYNA-N | |
10559 | |
CCC(C)I |
C4H9I | |
MFCD00001072 | |
sec-butyl iodide, butane, 2-iodo, 2-butyl iodide, sec-iodobutane, 2-jodbutan, 2-jodbutan czech, 2-iodanylbutane, 2-iodobutan, 2-iodo-butane, sec.-butyl iodide | |
2-iodobutane |
Specifications
513-48-4 | |
1.597 | |
21°C (70°F) | |
C4H9I | |
CH3CH2CHICH3 | |
50 g | |
1718777 | |
14,1574 | |
Insoluble in water. Soluble in alcohol and diethyl ether. | |
CCC(C)I | |
184.02 | |
184.02 | |
2-Iodobutane, stabilized with copper |
-104°C | |
119°C to 120°C | |
Ether-like | |
1.4991 | |
MFCD00001072 | |
UN2390 | |
Light sensitive | |
sec-butyl iodide, butane, 2-iodo, 2-butyl iodide, sec-iodobutane, 2-jodbutan, 2-jodbutan czech, 2-iodanylbutane, 2-iodobutan, 2-iodo-butane, sec.-butyl iodide | |
IQRUSQUYPCHEKN-UHFFFAOYNA-N | |
2-iodobutane | |
10559 | |
99% |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H302+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2-IODOBUTANE
EINECSNumber : 208-163-1
RTECSNumber : EK4410000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only