Learn More
2-Fluoro-5-(trifluoromethyl)phenol, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 141483-15-0 |
---|---|
Molecular Formula | C7H4F4O |
Molecular Weight (g/mol) | 180.10 |
MDL Number | MFCD00061286 |
InChI Key | MCOSBFKOUQAIJS-UHFFFAOYSA-N |
Synonym | 2-fluoro-5-trifluoromethyl phenol, 4-fluoro-3-hydroxybenzotrifluoride, 2-fluoro-5-trifluoromethyl-phenol, phenol, 2-fluoro-5-trifluoromethyl, alpha,alpha,alpha,6-tetrafluoro-m-cresol, pubchem4145, intermediates-zcf02079, acmc-1c6g6, ksc495m9p, 5-trifluoromethyl-2-fluorophenol |
PubChem CID | 518888 |
IUPAC Name | 2-fluoro-5-(trifluoromethyl)phenol |
SMILES | OC1=CC(=CC=C1F)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC397840010
|
Thermo Scientific Chemicals
397840010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
141483-15-0 | |
180.10 | |
MCOSBFKOUQAIJS-UHFFFAOYSA-N | |
518888 | |
OC1=CC(=CC=C1F)C(F)(F)F |
C7H4F4O | |
MFCD00061286 | |
2-fluoro-5-trifluoromethyl phenol, 4-fluoro-3-hydroxybenzotrifluoride, 2-fluoro-5-trifluoromethyl-phenol, phenol, 2-fluoro-5-trifluoromethyl, alpha,alpha,alpha,6-tetrafluoro-m-cresol, pubchem4145, intermediates-zcf02079, acmc-1c6g6, ksc495m9p, 5-trifluoromethyl-2-fluorophenol | |
2-fluoro-5-(trifluoromethyl)phenol |
Specifications
141483-15-0 | |
100.0 | |
146.0°C | |
Authentic | |
Glass bottle | |
1.4380 to 1.4400 (20°C, 589nm) | |
MFCD00061286 | |
1.436 | |
MCOSBFKOUQAIJS-UHFFFAOYSA-N | |
2-fluoro-5-(trifluoromethyl)phenol | |
518888 | |
98% |
97.5 | |
1.4360g/mL | |
57°C | |
97.5% min. (GC) | |
C7H4F4O | |
FC6H3(OH)CF3 | |
1g | |
2-fluoro-5-trifluoromethyl phenol, 4-fluoro-3-hydroxybenzotrifluoride, 2-fluoro-5-trifluoromethyl-phenol, phenol, 2-fluoro-5-trifluoromethyl, alpha,alpha,alpha,6-tetrafluoro-m-cresol, pubchem4145, intermediates-zcf02079, acmc-1c6g6, ksc495m9p, 5-trifluoromethyl-2-fluorophenol | |
OC1=CC(=CC=C1F)C(F)(F)F | |
180.10 | |
180.1 | |
2-Fluoro-5-(trifluoromethyl)phenol |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be con
GHS Signal Word: Warning
RUO â Research Use Only