Learn More
2-Chloro-6-methylphenyl isocyanate, 97%, Thermo Scientific Chemicals
$44.22 - $168.84
Chemical Identifiers
CAS | 40398-01-4 |
---|---|
Molecular Formula | C8H6ClNO |
Molecular Weight (g/mol) | 167.592 |
MDL Number | MFCD00037030 |
InChI Key | FBTQQNYGMICJQZ-UHFFFAOYSA-N |
Synonym | 2-chloro-6-methylphenyl isocyanate, 2-chloro-6-methylphenylisocyanate, benzene, 1-chloro-2-isocyanato-3-methyl, acmc-1ake5, 2-chloro-6-methylbenzenisocyanate, 2-chloro 6-methyl phenylisocyanate, 2-chloro 6-methyl phenyl isocyanate, 2-chloro-1-isocyanato-6-methyl benzene, 1-chloro-2-isocyanato-3-methyl-benzene, 6-chloro-2-methylphenylisocyanate |
PubChem CID | 268123 |
IUPAC Name | 1-chloro-2-isocyanato-3-methylbenzene |
SMILES | CC1=C(C(=CC=C1)Cl)N=C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1164503
|
Thermo Scientific Chemicals
L1164503 |
1 g |
Each for $44.22
|
|
|||||
AAL1164506
|
Thermo Scientific Chemicals
L1164506 |
5 g |
Each for $168.84
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
40398-01-4 | |
167.592 | |
FBTQQNYGMICJQZ-UHFFFAOYSA-N | |
268123 | |
CC1=C(C(=CC=C1)Cl)N=C=O |
C8H6ClNO | |
MFCD00037030 | |
2-chloro-6-methylphenyl isocyanate, 2-chloro-6-methylphenylisocyanate, benzene, 1-chloro-2-isocyanato-3-methyl, acmc-1ake5, 2-chloro-6-methylbenzenisocyanate, 2-chloro 6-methyl phenylisocyanate, 2-chloro 6-methyl phenyl isocyanate, 2-chloro-1-isocyanato-6-methyl benzene, 1-chloro-2-isocyanato-3-methyl-benzene, 6-chloro-2-methylphenylisocyanate | |
1-chloro-2-isocyanato-3-methylbenzene |
Specifications
40398-01-4 | |
79°C to 80°C (2 mmHg) | |
1.557 | |
MFCD00037030 | |
2690768 | |
FBTQQNYGMICJQZ-UHFFFAOYSA-N | |
1-chloro-2-isocyanato-3-methylbenzene | |
268123 | |
97% |
1.231 | |
1 g | |
C8H6ClNO | |
UN2206 | |
2-chloro-6-methylphenyl isocyanate, 2-chloro-6-methylphenylisocyanate, benzene, 1-chloro-2-isocyanato-3-methyl, acmc-1ake5, 2-chloro-6-methylbenzenisocyanate, 2-chloro 6-methyl phenylisocyanate, 2-chloro 6-methyl phenyl isocyanate, 2-chloro-1-isocyanato-6-methyl benzene, 1-chloro-2-isocyanato-3-methyl-benzene, 6-chloro-2-methylphenylisocyanate | |
CC1=C(C(=CC=C1)Cl)N=C=O | |
167.592 | |
167.6 | |
2-Chloro-6-methylphenyl isocyanate |
Safety and Handling
GHS H Statement
H330-H334-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P271-P280-P284-P285-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
H315-H319-H330-H334-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only