missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Butanone, +99%, Extra Pure, ACROS Organics™

$29.65 - $125.44

Chemical Identifiers

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.107
MDL Number MFCD00011648
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
Synonym 2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
PubChem CID 6569
ChEBI CHEBI:28398
IUPAC Name butan-2-one
SMILES CCC(=O)C
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC149670251 SDS View Documents Acros Organics
AC149670251
25L Metal drum N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC149670010 SDS View Documents Acros Organics
149670010
1L Glass Bottle Each for $70.69
AC149670025 SDS View Documents Acros Organics
149670025
2.5L Glass Bottle Each for $125.44
AC149670250 SDS View Documents Acros Organics
149670250
25mL Glass Bottle Each for $29.65
Specifications

Chemical Identifiers

78-93-3
72.107
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
6569
butan-2-one
C4H8O
MFCD00011648
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
CHEBI:28398
CCC(=O)C

Specifications

78-93-3
100.0
99+%
C2H5COCH3
01, IV, 3243
15, 6143
Solubility in water: 290g/L (20°C). Other solubilities: soluble in benzene, alcohol and ether, miscible with oils
CCC(=O)C
72.107
CHEBI:28398
Liquid
Extra Pure
Authentic
1.3780 to 1.3800
Solubility in water: 290g/L (20°C). Other solubilities: soluble in benzene, alcohol and ether, miscible with oils
0.2% max.
0.42 mPa.s (15°C)
Colorless
25L
99.0
Vapor Pressure: 105mbar at 20°C
C4H8O
MFCD00011648
01,678
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
butan-2-one
6569
72.11
≥99%
0.8060g/mL
Metal drum
0.005% max.
0.806
−7°C
80.0°C
-87.0°C
2-Butanone
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger