missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromoindene, 98%, Thermo Scientific Chemicals
$204.53 - $625.58
Chemical Identifiers
CAS | 10485-09-3 |
---|---|
Molecular Formula | C9H7Br |
Molecular Weight (g/mol) | 195.059 |
MDL Number | MFCD06797863 |
InChI Key | CCUYEVNCRQDQRF-UHFFFAOYSA-N |
Synonym | 2-bromoindene, 1h-indene, 2-bromo, indene, 2-bromo, 2-bromanyl-1h-indene, 2-bromoindene, 95+%, 2-bromo-indene, pubchem9657, #, acmc-1c5rn, sodium perborate,tetrahydrate |
PubChem CID | 575586 |
IUPAC Name | 2-bromo-1H-indene |
SMILES | C1C2=CC=CC=C2C=C1Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2547406
|
Thermo Scientific Chemicals
B2547406 |
5 g |
Each for $204.53
|
|
|||||
AAB2547414
|
Thermo Scientific Chemicals
B2547414 |
25 g |
Each for $625.58
|
|
|||||
Description
2-Bromoindene is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromoindene is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
10485-09-3 | |
195.059 | |
CCUYEVNCRQDQRF-UHFFFAOYSA-N | |
575586 | |
C1C2=CC=CC=C2C=C1Br |
C9H7Br | |
MFCD06797863 | |
2-bromoindene, 1h-indene, 2-bromo, indene, 2-bromo, 2-bromanyl-1h-indene, 2-bromoindene, 95+%, 2-bromo-indene, pubchem9657, #, acmc-1c5rn, sodium perborate,tetrahydrate | |
2-bromo-1H-indene |
Specifications
10485-09-3 | |
C9H7Br | |
5 g | |
Insoluble in water. | |
C1C2=CC=CC=C2C=C1Br | |
195.059 | |
195.06 | |
2-Bromoindene |
35°C to 37°C | |
MFCD06797863 | |
2-bromoindene, 1h-indene, 2-bromo, indene, 2-bromo, 2-bromanyl-1h-indene, 2-bromoindene, 95+%, 2-bromo-indene, pubchem9657, #, acmc-1c5rn, sodium perborate,tetrahydrate | |
CCUYEVNCRQDQRF-UHFFFAOYSA-N | |
2-bromo-1H-indene | |
575586 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
RUO – Research Use Only