Learn More
2-Bromoethanol, 97%, Thermo Scientific Chemicals
$63.79 - $835.18
Chemical Identifiers
CAS | 540-51-2 |
---|---|
Molecular Formula | C2H5BrO |
Molecular Weight (g/mol) | 124.965 |
MDL Number | MFCD00002827 |
InChI Key | LDLCZOVUSADOIV-UHFFFAOYSA-N |
Synonym | ethylene bromohydrin, ethanol, 2-bromo, 2-bromoethan-1-ol, glycol bromohydrin, 2-hydroxyethyl bromide, 2-bromo-1-ethanol, 1-bromo-2-ethanol, ethylenebromohydrin, 2-bromoethyl alcohol, 2-bromo-ethanol |
PubChem CID | 10898 |
IUPAC Name | 2-bromoethanol |
SMILES | C(CBr)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1027514
|
Thermo Scientific Chemicals
A1027514 |
25 g |
Each for $63.79
|
|
|||||
AAA1027530
|
Thermo Scientific Chemicals
A1027530 |
250 g |
Each for $254.05
|
|
|||||
AAA102750B
|
Thermo Scientific Chemicals
A102750B |
1000 g |
Each for $835.18
|
|
|||||
Description
2-Bromoethanol is utilized in selective reduction of nitroarenes. It is used as a solvent and organic synthetic raw material. It finds application in the synthesis of 2-piperidin-1-yl-ethanol by reacting with piperidine. It is used in selective reduction of nitroarenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromoethanol is utilized in selective reduction of nitroarenes. It is used as a solvent and organic synthetic raw material. It finds application in the synthesis of 2-piperidin-1-yl-ethanol by reacting with piperidine. It is used in selective reduction of nitroarenes.
Solubility
Miscible with water, organic solvent and petroleum ether.
Notes
Light sensitive. Hygroscopic. Store in a cool place. Incompatible with strong acids, strong oxidizing agents, strong reducing agents, acid chlorides and acid anhydrides.
Chemical Identifiers
540-51-2 | |
124.965 | |
LDLCZOVUSADOIV-UHFFFAOYSA-N | |
10898 | |
C(CBr)O |
C2H5BrO | |
MFCD00002827 | |
ethylene bromohydrin, ethanol, 2-bromo, 2-bromoethan-1-ol, glycol bromohydrin, 2-hydroxyethyl bromide, 2-bromo-1-ethanol, 1-bromo-2-ethanol, ethylenebromohydrin, 2-bromoethyl alcohol, 2-bromo-ethanol | |
2-bromoethanol |
Specifications
540-51-2 | |
1.762 | |
>110°C (230°F) | |
1.493 | |
MFCD00002827 | |
UN2922 | |
Hygroscopic; Light sensitive | |
ethylene bromohydrin, ethanol, 2-bromo, 2-bromoethan-1-ol, glycol bromohydrin, 2-hydroxyethyl bromide, 2-bromo-1-ethanol, 1-bromo-2-ethanol, ethylenebromohydrin, 2-bromoethyl alcohol, 2-bromo-ethanol | |
LDLCZOVUSADOIV-UHFFFAOYSA-N | |
2-bromoethanol | |
10898 | |
97% |
-80°C | |
149°C to 150°C | |
C2H5BrO | |
BrCH2CH2OH | |
25 g | |
878140 | |
14,3792 | |
Miscible with water,organic solvent and petroleum ether. | |
C(CBr)O | |
124.965 | |
124.97 | |
2-Bromoethanol |
Safety and Handling
GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P262-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301+H331-H310-H314
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 208-748-1
RTECSNumber : KJ8225000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only