missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Benzyloxybenzeneboronic acid, 96%, Thermo Scientific Chemicals
$82.92 - $269.26
Chemical Identifiers
CAS | 190661-29-1 |
---|---|
Molecular Formula | C13H13BO3 |
Molecular Weight (g/mol) | 228.054 |
MDL Number | MFCD01632206 |
InChI Key | MCAIDINWZOCYQK-UHFFFAOYSA-N |
Synonym | 2-benzyloxyphenylboronic acid, 2-benzyloxy phenylboronic acid, 2-benzyloxybenzeneboronic acid, 2-benzyloxy phenyl boronic acid, 2-benzyloxyphenyl boronic acid, 2-benzyloxyphenylborornic acid, 2-benzyloxy phenyl boranediol, o-benzyloxyphenylboronic acid, boronic acid, 2-phenylmethoxy phenyl |
PubChem CID | 2773253 |
IUPAC Name | (2-phenylmethoxyphenyl)boronic acid |
SMILES | B(C1=CC=CC=C1OCC2=CC=CC=C2)(O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL2010003
|
Thermo Scientific Chemicals
L2010003 |
1 g |
Each for $82.92
|
|
|||||
AAL2010006
|
Thermo Scientific Chemicals
L2010006 |
5 g |
Each for $269.26
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
190661-29-1 | |
228.054 | |
MCAIDINWZOCYQK-UHFFFAOYSA-N | |
2773253 | |
B(C1=CC=CC=C1OCC2=CC=CC=C2)(O)O |
C13H13BO3 | |
MFCD01632206 | |
2-benzyloxyphenylboronic acid, 2-benzyloxy phenylboronic acid, 2-benzyloxybenzeneboronic acid, 2-benzyloxy phenyl boronic acid, 2-benzyloxyphenyl boronic acid, 2-benzyloxyphenylborornic acid, 2-benzyloxy phenyl boranediol, o-benzyloxyphenylboronic acid, boronic acid, 2-phenylmethoxy phenyl | |
(2-phenylmethoxyphenyl)boronic acid |
Specifications
190661-29-1 | |
C13H13BO3 | |
1 g | |
2-benzyloxyphenylboronic acid, 2-benzyloxy phenylboronic acid, 2-benzyloxybenzeneboronic acid, 2-benzyloxy phenyl boronic acid, 2-benzyloxyphenyl boronic acid, 2-benzyloxyphenylborornic acid, 2-benzyloxy phenyl boranediol, o-benzyloxyphenylboronic acid, boronic acid, 2-phenylmethoxy phenyl | |
B(C1=CC=CC=C1OCC2=CC=CC=C2)(O)O | |
228.054 | |
228.06 | |
2-Benzyloxybenzeneboronic acid |
96°C to 99°C | |
MFCD01632206 | |
7813704 | |
MCAIDINWZOCYQK-UHFFFAOYSA-N | |
(2-phenylmethoxyphenyl)boronic acid | |
2773253 | |
96% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only