missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Amino-4-tert-butylthiazole, 98%, Thermo Scientific Chemicals
$56.42 - $617.05
Chemical Identifiers
CAS | 74370-93-7 |
---|---|
Molecular Formula | C7H12N2S |
Molecular Weight (g/mol) | 156.247 |
MDL Number | MFCD00051644 |
InChI Key | CUWZBHVYLVGOAB-UHFFFAOYSA-N |
Synonym | 2-amino-4-tert-butylthiazole, 4-tert-butylthiazol-2-ylamine, 4-tert-butyl thiazol-2-amine, 4-tert-butyl-thiazol-2-ylamine, 2-amino-4-t-butylthiazole, 4-tert-butyl-1,3-thiazol-2-amine, 4-tert-butylthiazol-2-amine, 4-tert-butyl-1,3-thiazol-2-ylamine, 2-thiazolamine, 4-1,1-dimethylethyl, 4-tert-butyl-1,3-thiazole-2-ylamine |
PubChem CID | 2734202 |
IUPAC Name | 4-tert-butyl-1,3-thiazol-2-amine |
SMILES | CC(C)(C)C1=CSC(=N1)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1411503
|
Thermo Scientific Chemicals
A1411503 |
1 g |
N/A
|
|
|||||
AAA1411506
|
Thermo Scientific Chemicals
A1411506 |
5 g |
N/A
|
|
|||||
AAA1411514
|
Thermo Scientific Chemicals
A1411514 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
74370-93-7 | |
156.247 | |
CUWZBHVYLVGOAB-UHFFFAOYSA-N | |
2734202 | |
CC(C)(C)C1=CSC(=N1)N |
C7H12N2S | |
MFCD00051644 | |
2-amino-4-tert-butylthiazole, 4-tert-butylthiazol-2-ylamine, 4-tert-butyl thiazol-2-amine, 4-tert-butyl-thiazol-2-ylamine, 2-amino-4-t-butylthiazole, 4-tert-butyl-1,3-thiazol-2-amine, 4-tert-butylthiazol-2-amine, 4-tert-butyl-1,3-thiazol-2-ylamine, 2-thiazolamine, 4-1,1-dimethylethyl, 4-tert-butyl-1,3-thiazole-2-ylamine | |
4-tert-butyl-1,3-thiazol-2-amine |
Specifications
74370-93-7 | |
C7H12N2S | |
1 g | |
2-amino-4-tert-butylthiazole, 4-tert-butylthiazol-2-ylamine, 4-tert-butyl thiazol-2-amine, 4-tert-butyl-thiazol-2-ylamine, 2-amino-4-t-butylthiazole, 4-tert-butyl-1,3-thiazol-2-amine, 4-tert-butylthiazol-2-amine, 4-tert-butyl-1,3-thiazol-2-ylamine, 2-thiazolamine, 4-1,1-dimethylethyl, 4-tert-butyl-1,3-thiazole-2-ylamine | |
CC(C)(C)C1=CSC(=N1)N | |
156.247 | |
156.26 | |
2-Amino-4-tert-butylthiazole |
98°C to 103°C | |
MFCD00051644 | |
114270 | |
CUWZBHVYLVGOAB-UHFFFAOYSA-N | |
4-tert-butyl-1,3-thiazol-2-amine | |
2734202 | |
98% |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
H302-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only