2,2'-Azino-bis-(3-ethyl-benzthiazoline-6-sulfonic Acid Solution, 100mL, MP Biomedicals
Stabilized Liquid Substrate A safe, convenient and good overall performing substrate for ELISA. It may be used for both kinetic and endpoint reactions; Supplied ready-to-use at 1.46 mmol/liter
Manufacturer:  MP Biochemicals LLC 04821809
Catalog No. ICN821809
Specifications
Specifications
2, 2'-Azino-bis-(3-ethyl-benzthiazoline-6-sulfonic Acid Solution, 100mL
ABTS
100 mL
Chemical Identifiers
ABTS
2, 2'-Azino-bis-(3-ethyl-benzthiazoline-6-sulfonic Acid Solution, 100mL
Documents