missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Phenoxy-2-propanol, tech. 85%, Thermo Scientific Chemicals

$67.26 - $218.12

Chemical Identifiers

CAS 770-35-4
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00016861
InChI Key IBLKWZIFZMJLFL-UHFFFAOYNA-N
Synonym 1-phenoxy-2-propanol, 2-propanol, 1-phenoxy, phenoxyisopropanol, propylene phenoxetol, 2-phenoxy-1-methylethanol, 1-phenoxy-propan-2-ol, phenyl-.beta.-hydroxypropyl ether, dsstox_cid_7312, dsstox_rid_78402, dsstox_gsid_27312
PubChem CID 92839
IUPAC Name 1-phenoxypropan-2-ol
SMILES CC(O)COC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0459618
View Documents Promotion Details
Thermo Scientific Chemicals
L0459618
50 g
Each for $67.26
 
AAL0459630
View Documents Promotion Details
Thermo Scientific Chemicals
L0459630
250 g
Each for $218.12
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

770-35-4
152.19
IBLKWZIFZMJLFL-UHFFFAOYNA-N
92839
CC(O)COC1=CC=CC=C1
C9H12O2
MFCD00016861
1-phenoxy-2-propanol, 2-propanol, 1-phenoxy, phenoxyisopropanol, propylene phenoxetol, 2-phenoxy-1-methylethanol, 1-phenoxy-propan-2-ol, phenyl-.beta.-hydroxypropyl ether, dsstox_cid_7312, dsstox_rid_78402, dsstox_gsid_27312
1-phenoxypropan-2-ol

Specifications

770-35-4
242°C to 244°C
C9H12O2
MFCD00016861
1941356
IBLKWZIFZMJLFL-UHFFFAOYNA-N
1-phenoxypropan-2-ol
92839
85%
1-Phenoxy-2-propanol
1.06
120°C (248°F)
1.523
50 g
1-phenoxy-2-propanol, 2-propanol, 1-phenoxy, phenoxyisopropanol, propylene phenoxetol, 2-phenoxy-1-methylethanol, 1-phenoxy-propan-2-ol, phenyl-.beta.-hydroxypropyl ether, dsstox_cid_7312, dsstox_rid_78402, dsstox_gsid_27312
CC(O)COC1=CC=CC=C1
152.19
152.19
Technical
Safety and Handling

Safety and Handling

EINECSNumber : 212-222-7

RTECSNumber : UB8886500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only