Learn More
1-Methylpyrrolidine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2379936
Description
1-Methylpyrrolidine is a methylated pyrollidine and is involved as an essential part of the structure of cefipime. It is also an active component of cigarette smoke.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Methylpyrrolidine is a methylated pyrollidine and is involved as an essential part of the structure of cefipime. It is also an active component of cigarette smoke.
Solubility
Miscible with water.
Notes
Air sensitive. Incompatible with strong oxidizing agents and strong acids.
Specifications
1-Methylpyrrolidine | |
-90°C | |
11.3 | |
−18°C (0°F) | |
Amine-like | |
1.4247 | |
500 g | |
n-methylpyrrolidine, methylpyrrolidine, n-methyltetrahydropyrrole, pyrrolidine, 1-methyl, n-methyl pyrrolidine, 1-methyl-pyrrolidine, pyrrolidine, n-methyl, unii-06509tzu6c, methyl pyrrolidine | |
AVFZOVWCLRSYKC-UHFFFAOYSA-N | |
1-methylpyrrolidine | |
8454 | |
98% |
120-94-5 | |
0.799 | |
80°C to 81°C | |
0.98 | |
C5H11N | |
MFCD00003173 | |
UN3286 | |
Miscible with water. | |
CN1CCCC1 | |
85.15 | |
85.15 |
Chemical Identifiers
120-94-5 | |
85.15 | |
AVFZOVWCLRSYKC-UHFFFAOYSA-N | |
8454 | |
CN1CCCC1 |
C5H11N | |
MFCD00003173 | |
n-methylpyrrolidine, methylpyrrolidine, n-methyltetrahydropyrrole, pyrrolidine, 1-methyl, n-methyl pyrrolidine, 1-methyl-pyrrolidine, pyrrolidine, n-methyl, unii-06509tzu6c, methyl pyrrolidine | |
1-methylpyrrolidine |
Safety and Handling
GHS H Statement
H225-H314-H318-H302-H332
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H301-H314-H332-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.
EINECSNumber : 204-438-5
RTECSNumber : UY1420500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only