Learn More
1-Methylimidazole, 99%, Thermo Scientific Chemicals
$71.18 - $620.51
Chemical Identifiers
CAS | 616-47-7 |
---|---|
Molecular Formula | C4H6N2 |
Molecular Weight (g/mol) | 82.11 |
MDL Number | MFCD00005292 |
InChI Key | MCTWTZJPVLRJOU-UHFFFAOYSA-N |
Synonym | 1-methyl-1h-imidazole, n-methylimidazole, 1h-imidazole, 1-methyl, imidazole, 1-methyl, n-methyl imidazole, 1-methyl-imidazole, unii-p4617qs63y, 1-methyl-1h-imidazol, n-methylimidazol, 1-methylimdazole |
PubChem CID | 1390 |
ChEBI | CHEBI:113454 |
SMILES | CN1C=CN=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1257522
|
Thermo Scientific Chemicals
A1257522 |
100 g |
Each for $71.18
|
|
|||||
AAA1257536
|
Thermo Scientific Chemicals
A1257536 |
500 g |
Each for $240.23
|
|
|||||
AAA12575A3
|
Thermo Scientific Chemicals
A12575A3 |
2 kg |
Each for $620.51
|
|
|||||
Description
1-Methylimidazole is used as a precursor for the synthesis of pyrrole-imidazole polyamides, ionic liquids such as 1-butyl-3-methylimidazolium hexafluorophosphate. It is actively involved in removing acid during the production of diethoxyphenylphosphine. It is used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Methylimidazole is used as a precursor for the synthesis of pyrrole-imidazole polyamides, ionic liquids such as 1-butyl-3-methylimidazolium hexafluorophosphate. It is actively involved in removing acid during the production of diethoxyphenylphosphine. It is used as an intermediate in organic synthesis.
Solubility
Miscible with water.
Notes
Incompatible with carbon dioxide and strong oxidizing agents.
Chemical Identifiers
616-47-7 | |
82.11 | |
MCTWTZJPVLRJOU-UHFFFAOYSA-N | |
1390 | |
CN1C=CN=C1 |
C4H6N2 | |
MFCD00005292 | |
1-methyl-1h-imidazole, n-methylimidazole, 1h-imidazole, 1-methyl, imidazole, 1-methyl, n-methyl imidazole, 1-methyl-imidazole, unii-p4617qs63y, 1-methyl-1h-imidazol, n-methylimidazol, 1-methylimdazole | |
CHEBI:113454 |
Specifications
616-47-7 | |
1.031 | |
92°C (197°F) | |
1.497 | |
100 g | |
105197 | |
1-methyl-1h-imidazole, n-methylimidazole, 1h-imidazole, 1-methyl, imidazole, 1-methyl, n-methyl imidazole, 1-methyl-imidazole, unii-p4617qs63y, 1-methyl-1h-imidazol, n-methylimidazol, 1-methylimdazole | |
MCTWTZJPVLRJOU-UHFFFAOYSA-N | |
1-methyl-1H-imidazole | |
1390 | |
82.11 | |
1-Methylimidazole |
-6°C | |
195°C to 197°C | |
C4H6N2 | |
MFCD00005292 | |
UN3267 | |
Hygroscopic | |
Miscible with water. | |
CN1C=CN=C1 | |
82.11 | |
CHEBI:113454 | |
99% |
Safety and Handling
GHS H Statement
H314-H302-H312-H227
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302-H311-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 210-484-7
RTECSNumber : NI7000000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only