Learn More
1-Methoxy-2-propanol, 98.5%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 244990025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Methoxy-2-propanol | |
-97.0°C | |
120.0°C | |
Authentic | |
Glass Bottle | |
1.4020 to 1.4040 | |
MFCD00004537 | |
01,536 | |
0.92 | |
Solubility in water: 1000g/L (20°C) | |
COCC(C)O | |
90.12 | |
1.7 mPa.s (20°C) | |
98.5% | |
Liquid |
107-98-2 | |
0.9200g/mL | |
30°C | |
98% min. (GC) | |
C4H10O2 | |
CH3OCH2CH(OH)CH3 | |
2.5 L | |
0.01% max. | |
1-methoxy-2-propanol, pgme, methoxyisopropanol, 2-propanol, 1-methoxy, closol, dowtherm 209, propylene glycol monomethyl ether, poly-solve mpm, propasol solvent m, dowanol 33b | |
ARXJGSRGQADJSQ-UHFFFAOYNA-N | |
1-methoxypropan-2-ol | |
7900 | |
90.12 | |
Extra Pure |
Chemical Identifiers
107-98-2 | |
90.12 | |
ARXJGSRGQADJSQ-UHFFFAOYNA-N | |
7900 | |
COCC(C)O |
C4H10O2 | |
MFCD00004537 | |
1-methoxy-2-propanol, pgme, methoxyisopropanol, 2-propanol, 1-methoxy, closol, dowtherm 209, propylene glycol monomethyl ether, poly-solve mpm, propasol solvent m, dowanol 33b | |
1-methoxypropan-2-ol |
Safety and Handling
GHS H Statement
May cause drowsiness or dizziness.
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest
GHS Signal Word: Danger
EINECSNumber : 203-539-1
RUO – Research Use Only