missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (1-Hexadecyl)pyridinium chloride monohydrate, 98%

$17.50 - $154.08

Chemical Identifiers

CAS 6004-24-6
Molecular Formula C21H40ClNO
Molecular Weight (g/mol) 358.007
MDL Number MFCD00149977
InChI Key NFCRBQADEGXVDL-UHFFFAOYSA-M
Synonym cetylpyridinium chloride monohydrate, 1-hexadecylpyridin-1-ium chloride hydrate, 1-hexadecylpyridinium chloride monohydrate, hexadecylpyridinium chloride monohydrate, unii-d9om4sk49p, pyridinium, 1-hexadecyl-, chloride, monohydrate, cetylpyridinium chloride monohydrate, d9om4sk49p, cetylpyridinii chloridum, sprol tn
PubChem CID 22324
ChEBI CHEBI:3566
IUPAC Name 1-hexadecylpyridin-1-ium;chloride;hydrate
SMILES CCCCCCCCCCCCCCCC[N+]1=CC=CC=C1.O.[Cl-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1349922 SDS View Documents Alfa Aesar™
A1349922
100g Each for $46.38
AAA1349936 SDS View Documents Alfa Aesar™
A1349936
500g Each for $154.08
AAA1349914 SDS View Documents Alfa Aesar™
A1349914
25g Each for $17.50
Description

Description

Applications
(1-Hexadecyl)pyridinium chloride monohydrate is used in a study to assess removal of emulsified oil from oily waste water and removal of emulsified food and mineral oils from waste water. And it is used for detergent-based extraction of membrane-associated enzymes such as alkaline phosphatase, and in extraction of chromium and palladium.

Solubility
Soluble in water, alcohol, chloroform

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

6004-24-6
358.007
NFCRBQADEGXVDL-UHFFFAOYSA-M
22324
1-hexadecylpyridin-1-ium;chloride;hydrate
C21H40ClNO
MFCD00149977
cetylpyridinium chloride monohydrate, 1-hexadecylpyridin-1-ium chloride hydrate, 1-hexadecylpyridinium chloride monohydrate, hexadecylpyridinium chloride monohydrate, unii-d9om4sk49p, pyridinium, 1-hexadecyl-, chloride, monohydrate, cetylpyridinium chloride monohydrate, d9om4sk49p, cetylpyridinii chloridum, sprol tn
CHEBI:3566
CCCCCCCCCCCCCCCC[N+]1=CC=CC=C1.O.[Cl-]

Specifications

83°C to 86°C
6004-24-6
MFCD00149977
3578606
cetylpyridinium chloride monohydrate, 1-hexadecylpyridin-1-ium chloride hydrate, 1-hexadecylpyridinium chloride monohydrate, hexadecylpyridinium chloride monohydrate, unii-d9om4sk49p, pyridinium, 1-hexadecyl-, chloride, monohydrate, cetylpyridinium chloride monohydrate, d9om4sk49p, cetylpyridinii chloridum, sprol tn
NFCRBQADEGXVDL-UHFFFAOYSA-M
1-hexadecylpyridin-1-ium;chloride;hydrate
22324
358.02 (339.99 Anhydrous)
(1-Hexadecyl)pyridinium chloride monohydrate
100g
C21H40ClNO
UN2811
14,2032
Soluble in water, alcohol, chloroform
CCCCCCCCCCCCCCCC[N+]1=CC=CC=C1.O.[Cl-]
358.007
CHEBI:3566
98%
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : UU5075000

TSCA : TSCA