Learn More
(1-Hexadecyl)pyridinium chloride monohydrate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1349936
Description
(1-Hexadecyl)pyridinium chloride monohydrate is used in a study to assess removal of emulsified oil from oily waste water and removal of emulsified food and mineral oils from waste water. And it is used for detergent-based extraction of membrane-associated enzymes such as alkaline phosphatase, and in extraction of chromium and palladium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(1-Hexadecyl)pyridinium chloride monohydrate is used in a study to assess removal of emulsified oil from oily waste water and removal of emulsified food and mineral oils from waste water. And it is used for detergent-based extraction of membrane-associated enzymes such as alkaline phosphatase, and in extraction of chromium and palladium.
Solubility
Soluble in water, alcohol, chloroform
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
(1-Hexadecyl)pyridinium chloride monohydrate | |
83°C to 86°C | |
MFCD00149977 | |
UN2811 | |
14,2032 | |
Soluble in water, alcohol, chloroform | |
O.[Cl-].CCCCCCCCCCCCCCCCN1C=C[CH+]C=C1 | |
358.01 | |
CHEBI:3566 | |
98% |
6004-24-6 | |
C21H40ClNO | |
500 g | |
3578606 | |
cetylpyridinium chloride monohydrate, 1-hexadecylpyridin-1-ium chloride hydrate, 1-hexadecylpyridinium chloride monohydrate, hexadecylpyridinium chloride monohydrate, unii-d9om4sk49p, pyridinium, 1-hexadecyl-, chloride, monohydrate, d9om4sk49p, cetylpyridinii chloridum, sprol tn | |
NFCRBQADEGXVDL-UHFFFAOYSA-M | |
1-hexadecyl-1,4-dihydropyridin-4-ylium hydrate chloride | |
22324 | |
358.02 (339.99 Anhydrous) |
Chemical Identifiers
6004-24-6 | |
358.01 | |
NFCRBQADEGXVDL-UHFFFAOYSA-M | |
22324 | |
O.[Cl-].CCCCCCCCCCCCCCCCN1C=C[CH+]C=C1 |
C21H40ClNO | |
MFCD00149977 | |
cetylpyridinium chloride monohydrate, 1-hexadecylpyridin-1-ium chloride hydrate, 1-hexadecylpyridinium chloride monohydrate, hexadecylpyridinium chloride monohydrate, unii-d9om4sk49p, pyridinium, 1-hexadecyl-, chloride, monohydrate, d9om4sk49p, cetylpyridinii chloridum, sprol tn | |
CHEBI:3566 |
Safety and Handling
GHS H Statement
H300-H330-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H301-H315-H318-H330-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 204-593-9
RTECSNumber : UU5075000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only