Learn More
1-Fluoro-2-iodobenzene, 99%, stab. with copper, Thermo Scientific Chemicals
$66.56 - $889.75
Chemical Identifiers
CAS | 348-52-7 |
---|---|
Molecular Formula | C6H4FI |
Molecular Weight (g/mol) | 222.00 |
MDL Number | MFCD00001031 |
InChI Key | TYHUGKGZNOULKD-UHFFFAOYSA-N |
Synonym | 2-fluoroiodobenzene, o-fluoroiodobenzene, o-iodofluorobenzene, benzene, 1-fluoro-2-iodo, 2-iodofluorobenzene, 3-fluoro-2-iodobenzene, 1-fluoro-2-iodo-benzene, 2-fluoro-iodobenzene, 2-fluoro-1-iodobenzene |
PubChem CID | 67673 |
IUPAC Name | 1-fluoro-2-iodobenzene |
SMILES | FC1=CC=CC=C1I |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1241114
|
Thermo Scientific Chemicals
A1241114 |
25 g |
N/A
|
|
|||||
AAA1241122
|
Thermo Scientific Chemicals
A1241122 |
100 g |
N/A
|
|
|||||
AAA1241136
|
Thermo Scientific Chemicals
A1241136 |
500 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
348-52-7 | |
222.00 | |
TYHUGKGZNOULKD-UHFFFAOYSA-N | |
67673 | |
FC1=CC=CC=C1I |
C6H4FI | |
MFCD00001031 | |
2-fluoroiodobenzene, o-fluoroiodobenzene, o-iodofluorobenzene, benzene, 1-fluoro-2-iodo, 2-iodofluorobenzene, 3-fluoro-2-iodobenzene, 1-fluoro-2-iodo-benzene, 2-fluoro-iodobenzene, 2-fluoro-1-iodobenzene | |
1-fluoro-2-iodobenzene |
Specifications
348-52-7 | |
1.903 | |
71°C (159°F) | |
1.59 | |
25 g | |
Light sensitive | |
TYHUGKGZNOULKD-UHFFFAOYSA-N | |
1-fluoro-2-iodobenzene | |
67673 | |
99% |
-43°C to -40°C | |
188°C to 189°C | |
C6H4FI | |
MFCD00001031 | |
2039770 | |
2-fluoroiodobenzene, o-fluoroiodobenzene, o-iodofluorobenzene, benzene, 1-fluoro-2-iodo, 2-iodofluorobenzene, 3-fluoro-2-iodobenzene, 1-fluoro-2-iodo-benzene, 2-fluoro-iodobenzene, 2-fluoro-1-iodobenzene | |
FC1=CC=CC=C1I | |
222.00 | |
222 | |
1-Fluoro-2-iodobenzene, Stabilized with copper |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302-H315-H319-H335
EINECSNumber : 206-477-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only