Learn More
1-Butanol, HPLC Grade, 99%, Thermo Scientific Chemicals
Filtered through 0.2μm filters
$114.70 - $347.79
Chemical Identifiers
CAS | 71-36-3 |
---|---|
Molecular Formula | C4H10O |
Molecular Weight (g/mol) | 74.123 |
MDL Number | MFCD00002964 |
InChI Key | LRHPLDYGYMQRHN-UHFFFAOYSA-N |
Synonym | 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane |
PubChem CID | 263 |
ChEBI | CHEBI:28885 |
IUPAC Name | butan-1-ol |
SMILES | CCCCO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA22925K2
|
Thermo Scientific Chemicals
022925K2 |
1 L |
Each for $114.70
|
|
|||||
AA22925M1
|
Thermo Scientific Chemicals
022925M1 |
4 x 1 L |
Each for $347.79
|
|
|||||
AA22925K7
|
Thermo Scientific Chemicals
022925K7 |
4 L |
Each for $281.23
|
|
|||||
Description
1-Butanol is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
71-36-3 | |
74.123 | |
LRHPLDYGYMQRHN-UHFFFAOYSA-N | |
263 | |
butan-1-ol |
C4H10O | |
MFCD00002964 | |
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane | |
CHEBI:28885 | |
CCCCO |
Specifications
−90°C to -89°C | |
0.810 | |
1 L | |
Liquid | |
99% | |
MFCD00002964 | |
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane | |
LRHPLDYGYMQRHN-UHFFFAOYSA-N | |
butan-1-ol | |
263 | |
74.12 | |
1-Butanol |
71-36-3 | |
117°C to 118°C | |
35°C (95°F) | |
1.399 | |
C4H10O | |
969148 | |
Miscible with alcohols,ketones,aldehydes,ethers,aliphatic and aromatic hydrocarbons. Slightly miscible with water. | |
CCCCO | |
74.123 | |
CHEBI:28885 | |
HPLC |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P363-P370+P378q-P501c
H226-H302-H315-H318-H335-H336
DOTInformation : Hazard Class: 3; Packaging Group: III
EINECSNumber : 200-751-6
RTECSNumber : EO1400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only