Learn More
1-Adamantylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H58408AD
Description
1-Adamantylzinc bromide is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Adamantylzinc bromide is used as pharmaceutical intermediates.
Solubility
Reacts with water.
Notes
Air Sensitive. Incompatible with oxidizing agents, bases and heat. Store under dry inert gas. Sto
Specifications
1-Adamantylzinc bromide | |
312624-15-0 | |
C10H15BrZn | |
50 mL | |
Air sensitive; May form a precipitate | |
Reacts with water. | |
C1C2CC3CC1C[C-](C2)C3.[Zn+]Br | |
280.514 | |
280.51 |
Packaged under Argon in resealable ChemSeal™ bottles | |
0.97 | |
MFCD00671988 | |
UN3399 | |
1-adamantylzinc bromide, 1-adamantylzinc bromide solution, adamantan-1-yl bromo zinc, 1-adamantylzinc bromide solution, 0.5 m in thf, bromo 3r,5s,7s-adamantan-1-yl zinc, 1-adamantylzinc bromide 0.5 m in tetrahydrofuran, 1-adamantylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles | |
PTWVHBHYYPLVCD-UHFFFAOYSA-M | |
adamantan-1-ide;bromozinc(1+) | |
5141693 |
Chemical Identifiers
312624-15-0 | |
280.514 | |
PTWVHBHYYPLVCD-UHFFFAOYSA-M | |
5141693 | |
C1C2CC3CC1C[C-](C2)C3.[Zn+]Br |
C10H15BrZn | |
MFCD00671988 | |
1-adamantylzinc bromide, 1-adamantylzinc bromide solution, adamantan-1-yl bromo zinc, 1-adamantylzinc bromide solution, 0.5 m in thf, bromo 3r,5s,7s-adamantan-1-yl zinc, 1-adamantylzinc bromide 0.5 m in tetrahydrofuran, 1-adamantylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles | |
adamantan-1-ide;bromozinc(1+) |
Safety and Handling
GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
H225-H260-H302-H314-H335-H336-H351
DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only