Learn More
1-(2-Aminoethyl)piperidine, 98%, Thermo Scientific Chemicals
$82.92 - $264.19
Chemical Identifiers
CAS | 27578-60-5 |
---|---|
Molecular Formula | C7H16N2 |
Molecular Weight (g/mol) | 128.219 |
MDL Number | MFCD00006516 |
InChI Key | CJNRGSHEMCMUOE-UHFFFAOYSA-N |
Synonym | n-2-aminoethyl piperidine, 1-2-aminoethyl piperidine, 2-piperidin-1-yl ethanamine, 2-piperidin-1-yl ethan-1-amine, 1-piperidineethanamine, 2-piperidinoethylamine, 2-1-piperidinyl ethanamine, 2-1-piperidinyl ethylamine, n-aminoethylpiperidine, 2-piperidino-1-ethanamine |
PubChem CID | 33944 |
IUPAC Name | 2-piperidin-1-ylethanamine |
SMILES | C1CCN(CC1)CCN |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1837306
|
Thermo Scientific Chemicals
A1837306 |
5 g |
Each for $82.92
|
|
|||||
AAA1837314
|
Thermo Scientific Chemicals
A1837314 |
25 g |
Each for $264.19
|
|
|||||
Description
1-(2-Aminoethyl)piperidine is used in modification of vinylbenzyl chloride/divinylbenzene gel copolymer bead and in the synthesis of linkage isomers trans-bis[1-(2-aminoethyl)piperidine]dinitronickel and trans-bis[1-(2-aminoethyl)-piperidine]dinitritonickel. It is also used as a reactant for synthesis of: analogs of anticancer agents, inhibitor of botulinum neurotoxin serotype A light chain, P. falciparum malaria, and Ebola filovirus, cannabinoid CB1 receptor antagonists, small molecules that restore E-cadherin expression and reduce invasion in colorectal carcinoma cells, potent and selective 5-HT6 antagonists and N-mustards as anticancer agents. 1-(2-Aminoethyl)piperidine on [1+1] condensation reaction with 3-methoxy salicylaldehyde yields tridentate Schiff base ligand.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-(2-Aminoethyl)piperidine is used in modification of vinylbenzyl chloride/divinylbenzene gel copolymer bead and in the synthesis of linkage isomers trans-bis[1-(2-aminoethyl)piperidine]dinitronickel and trans-bis[1-(2-aminoethyl)-piperidine]dinitritonickel. It is also used as a reactant for synthesis of: analogs of anticancer agents, inhibitor of botulinum neurotoxin serotype A light chain, P. falciparum malaria, and Ebola filovirus, cannabinoid CB1 receptor antagonists, small molecules that restore E-cadherin expression and reduce invasion in colorectal carcinoma cells, potent and selective 5-HT6 antagonists and N-mustards as anticancer agents. 1-(2-Aminoethyl)piperidine on [1+1] condensation reaction with 3-methoxy salicylaldehyde yields tridentate Schiff base ligand.
Solubility
Partly miscible in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, acid chlorides and acid anhydrides. It is sensitive to air.
Chemical Identifiers
27578-60-5 | |
128.219 | |
CJNRGSHEMCMUOE-UHFFFAOYSA-N | |
33944 | |
C1CCN(CC1)CCN |
C7H16N2 | |
MFCD00006516 | |
n-2-aminoethyl piperidine, 1-2-aminoethyl piperidine, 2-piperidin-1-yl ethanamine, 2-piperidin-1-yl ethan-1-amine, 1-piperidineethanamine, 2-piperidinoethylamine, 2-1-piperidinyl ethanamine, 2-1-piperidinyl ethylamine, n-aminoethylpiperidine, 2-piperidino-1-ethanamine | |
2-piperidin-1-ylethanamine |
Specifications
27578-60-5 | |
185°C to 186°C | |
C7H16N2 | |
MFCD00006516 | |
UN2733 | |
Air Sensitive | |
Partly miscible in water. | |
C1CCN(CC1)CCN | |
128.219 | |
128.22 | |
1-(2-Aminoethyl)piperidine |
0.903 | |
57°C (134°F) | |
1.474 | |
5 g | |
103469 | |
n-2-aminoethyl piperidine, 1-2-aminoethyl piperidine, 2-piperidin-1-yl ethanamine, 2-piperidin-1-yl ethan-1-amine, 1-piperidineethanamine, 2-piperidinoethylamine, 2-1-piperidinyl ethanamine, 2-1-piperidinyl ethylamine, n-aminoethylpiperidine, 2-piperidino-1-ethanamine | |
CJNRGSHEMCMUOE-UHFFFAOYSA-N | |
2-piperidin-1-ylethanamine | |
33944 | |
98% |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber : 248-540-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only