Fisher BioReagents FASTRun Tris SDS PAGE Running Buffer
Fisher BioReagents FASTRun Tris SDS PAGE Running Buffer Fisher BioReagents FASTRun Tris SDS PAGE Running Buffer
With 10x FASTRun Tris SDS PAGE Running Buffer you can significantly reduce the time you spend waiting for gels while seeing comparable or better resolution than with traditional buffers.
  • Compatible with standard polyacrylamide gels (whether you buy them premade or pour your own)
  • Compatible with all commercially available protein electrophoresis tanks
A New and Faster Way to Separate Protein Gels
Fisher BioReagents FASTRun Tris SDS PAGE Running Buffer provides clear results in significantly less time than traditional buffers.
2868-LDGEN-GC-FastRun-Buffer-IMAGE
Image 1: Traditional Tris-Glycine SDS, 125V, 25 minutes
Image 2: FASTRun Buffer, 200V, 20 minutes
Image 3: Traditional Tris-Glycine SDS, 125V, 90 minutes