Cell Culture Media
HyClone™ Dulbecco's High Glucose Modified Eagles Medium
Designed to support cell culture growth and biomanufacturing processes.
HyClone™ RPMI 1640 Media
Designed to support cell culture growth and biomanufacturing processes.
HyClone™ Dulbecco's Modified Eagles Medium
Designed to support cell culture growth and biomanufacturing processes.
Gibco™ DMEM, High Glucose DFS Item
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, High Glucose is used in cell culture processing applications.
Corning™ DMEM with L-Glutamine, 4.5g/L Glucose and Sodium Pyruvate
Subsititution of Pyridoxine HCl for Pyridoxal ensures improved stability over original formulation. Corning™ DMEM with L-Glutamine, 4.5g/L Glucose and Sodium Pyruvate is hybridoma tested.
Gibco™ Neurobasal™ Medium DFS Item
Used in the growth of neuronal cells from hippocampus, cortex and otregions of the brain. Gibco™ Neurobasal™ Medium is a basal medium that meets the special cell culture requirements of pre-natal and embryonic neuronal cells when used with Gibco™ B-27™ Supplement.
Gibco™ Recovery™ Cell Culture Freezing Medium, DMSO (10%) DFS Item
Complete, ready-to-use freezing medium for cryopreservation of a wide variety of mammalian cells, including CHO-S, CHO-K1, HEK 293, Jurkat, and NIH 3T3
Gibco™ Opti-MEM™ I Reduced Serum Medium DFS Item
An improved Minimal Essential Medium that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in growth rate or morphology
Gibco™ DMEM, High Glucose, Pyruvate DFS Item
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, High Glucose, Pyruvate is used in cell culture processing applications.
Gibco™ RPMI 1640 Medium DFS Item
Originally developed to culture human leukemic cells in suspension and as a monolayer
Gibco™ DMEM, No Glucose DFS Item
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, No Glucose is used in cell culture processing applications.
Gibco™ Advanced DMEM/F-12 DFS Item
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ Advanced DMEMF/12 is used in cell culture processing applications.
HyClone™ Minimum Essential Medium
Support cell culture growth and biomanufacturing processes with HyClone™ Minimum Essential Medium (MEM) with Earle's Balanced Salts (MEM/EBSS), manufactured based on compliance with ISO 9001:2000 guidelines and current Good Manufacturing Practices (cGMP) for Medical Devices.
Gibco™ Human Endothelial-SFM DFS Item
Human Endothelial-SFM
Gibco™ Keratinocyte-SFM (1X) DFS Item
Keratinocyte-SFM (1X)
HyClone™ DMEM/F12 1:1 Media
Used for culturing many different mammalian cells including MDCK, Glial cells, fibroblasts, and endothelial cells. HyClone™ DMEM/F12 1:1 Media supports cell culture growth and biomanufacturing processes and is manufactured using ISO 9001:2000-certified processes.
HyClone™ Iscove's Modified Dulbecco's Medium (IMDM)
Support cell culture growth and biomanufacturing processes with HyClone™ Iscove's Modified Dulbecco's Medium (IMDM), manufactured using ISO 9001:2000-certified processes.
Gibco™ RPMI 1640 Medium, No Phenol Red DFS Item
Originally developed to culture human leukemic cells in suspension and as a monolayer
Gibco™ DMEM/F-12, HEPES DFS Item
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEMF/12, HEPES is used in cell culture processing applications.
HyClone™ Leibovitz L-15 Media
Designed for culture without CO2 equilibration.
Gibco™ Opti-MEM™ I Reduced Serum Medium, No Phenol Red DFS Item
Improved Minimal Essential Medium that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in cell growth rate or morphology
Gibco™ Leibovitz's L-15 Medium DFS Item
Supportsgrowth of HEP-2 monkey kidney cells and primary explants of embryonic and adult human tissue
Corning™ RPMI 1640 Medium (Mod.) 1X with L-Glutamine
Guaranteed low endotoxin levels ≤0.025EU/mL
Gibco™ RPMI 1640 Medium, No Glucose DFS Item
Originally developed to culture human leukemic cells in suspension and as a monolayer
Gibco™ RPMI 1640 Medium (ATCC Modification), HEPES, Liquid DFS Item
Originally developed to culture human leukemic cells in suspension and as a monolayer
Gibco™ MEM DFS Item
Minimum Essential Medium is one of the most commonly used of all cell culture media
Gibco™ Collagenase, Type IV, Powder DFS Item
Cleaves bond between neutral amino acid (X) and glycine in sequence Pro-X-Glyc-Pro, which is found with high frequency in collagen
Corning™ DMSO (Dimethyl Sulfoxide)
For cryopreservation of cells along with glycerol, to prevent damage to cell membrane during freezing
  spinner