Funnels
Thermo Scientific™ Nalgene™ Polypropylene Powder Funnels
Transfer powders with these powder funnels. Parallel stem minimizes bridging of powder and external ribbing prevents air locks. A shatter-proof alternative to glass.
Thermo Scientific™ Nalgene™ Analytical Test Filter Funnels
Perform microbiological QC testing and analysis of water, food/beverage, raw material and finished product with these economical sterile disposable funnels.
Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels
Use standard filter papers with Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels, featuring long stems and external ribbing that eliminates air lock.
Fisherbrand™ Reusable Glass Short-Stem Funnels
High resistance to chemical attack, mechanical, and thermal shock
Fisherbrand™ Powder Funnels
Parallel stems minimize bridging of powder
Fisherbrand™ Weighing Funnels
Weighing dish and sample transfer funnel in one.
DWK Life Sciences Kimble™ KIMAX™ Filling Funnels
Fire-polished edges reduce chipping
United Scientific Supplies Separatory Funnels
Provide excellent clarity and strength.
Bel-Art™ SP Scienceware™ Buchner Funnels
Easy-to-clean two-piece, polypropylene with perforated HDPE plastic filter plate
DWK Life Sciences Kimble™ KIMAX™ Long Stem 60° Angle Addition Funnels
High resistance to chemical attack, mechanical and thermal shock
Medegen Stainless-Steel Funnels
Noncorrosive 304 stainless steel
DWK Life Sciences Kimble™ Kontes™ Parts for Microfiltration Assemblies
For use with ULTRA-WARE™ microfiltration assembly
Bel-Art™ SP Scienceware™ Polypropylene Powder Funnels
Feature a large bore stem to minimize bridging of powder
Thermo Scientific™ Nalgene™ Reusable Filter Funnel with Clamp
Perform high throughput particle and microbiological QC testing using this reusable filter funnel. Quick release clamp provides fast and easy retrieval of membranes.
TWD Scientific™ Tradewinds Direct Weighing Funnels
Disposable polypropylene funnels in four different sizes
Fisherbrand™ Glass Funnel
Provides for easy buret filling.
PYREX™ Funnel for Sugar Analysis
Molded construction with bottom ground flat and beaded rim
Thermo Scientific™ Nalgene™ Polysulfone Reusable Bottle Top Filters
Combine the convenience of a bottle top filter with the savings of a reusable bottle top filter. Screw securely onto glass media bottles with 33 or 45mm neck sizes.
PYREX™ Funnels with Fluted Bowls and Short Stems
Useful in clinical and hospital labs; can be used in flasks or tubes without racks
Bel-Art™ SP Scienceware™ Polypropylene Funnels with Short Stems and External Ridges
Sturdy, polypropylene funnels with ridges help prevent vapor lock
EMD Millipore™ Funnel and Locking Ring Assembly
Analyze particulate or biological contamination via vacuum filtration
PYREX™ Fluted Funnel
Useful in clinical and hospital labs; can be used in flasks or tubes without racks.
Bel-Art™ SP Scienceware™ Polypropylene Long-Stemmed Funnels
Heavy-duty design with external ribs to prevent air lock
PYREX™ Squibb Separatory Funnel, PTFE Product Standard Stopcock, Standard Taper Joints
With No. 2 Product Standard stopcocks and inner and outer 19/22 Standard Taper joints.
Thermo Scientific™ Nalgene™ Reusable Filter Holders with Receiver
Eliminate the need for a manifold or breakable glass vacuum flask. Nalgene Reusable Filter Holders with Receiver are complete, environmentally-friendly systems.
  spinner